Versaflo™ M-Series Respiratory Faceshield Assembly, 3M™
Catalog # 10837-154
Supplier: 3M
Specifications
- Color:Gray
- Description:Ratchet Suspension with Flame-Resistant Faceseal
- Size:Standard
- Environmentally Preferable:
- Cat. No.:10837-154
- Supplier no.:51141567402
Specifications
About this item
M-Series is a lightweight, comfortable PAPR headtop that combines a respirator and faceshield, with the option of adding earmuffs for hearing protection.
This headtop is popular in light industry, where respirators and face protection are typically used.
Frequently Bought Together