ContactUsLinkComponent
Feather Double Edge Blade
Feather Double Edge Blade, Electron Microscopy Sciences
Catalog # 101413-472
Minority or Woman-Owned Business Enterprise
undefined
Feather Double Edge Blade, Electron Microscopy Sciences
Catalog # 101413-472
Supplier Number:  72002-01
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Feather Double Edge Blade
  • Environmentally Preferable:
  • Cat. No.:
    101413-472
  • Supplier no.:
    72002-01
Specifications
About this item
Made from carbon steel.
Feather® blades are well known to have super sharp edges. Easily breaks in half for use with any tissue slicing machine, or break smaller portions for use as a microscalpel with a Blade Breaker and Holder. Blades are available in packages of 10 blades per box or 500 blades per box. Measurements W x L x T: 21.95 x 42.96 x 0.10 mm (0.865 x 1.69 x 0.004")
Frequently Bought Together