About this item
Enables development of more efficient plant transformation systems.
Requires supplemental thymidine for growth
- Reduced bacterial overgrowth during co-cultivation
- Decreased need for antibiotics
Intact Genomics (ig®) Thymidine Auxotrophic EHA105Thy Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless 50 mg/L of thymidine is added to minimal medium.
This prevents the bacteria from overgrowing plant tissues when used for plant transformation.
Thymidine-Auxotrophic EHA105Thy Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies.
The EHA105Thy strain is useful for transgenic operations of rice, tobacco and other plants. EAH105Thy– thyA knockout mutant derived from EHA105, itself a derivative of A281 (A136/pTiBo542).
Manufactured under ISO 13485:2016 certified quality management system.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 104090-482
- 104090-484
- Storage Conditions:
- –80 °C
- –80 °C