Anti-TAF5L Rabbit Polyclonal Antibody
Catalog # 77806-911
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Synthesized peptide derived from internal of TAF5L
- Antigen Symbol:TAF5L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O75529
- Antigen Synonyms:1110005N04Rik|AI849020|OTTHUMP00000037470|OTTHUMP00000037471|PAF65 beta|PAF65-beta|PAF65B|PCAF associated factor 65 beta|PCAF-associated factor 65 beta|TAF 5L|TAF5 like RNA polymerase II p300/CBP associated factor (PCAF) associated factor 65 kD|TAF5 like RNA polymerase II p300/CBP associated factor (PCAF) associated factor 65 kDa|TAF5 like RNA polymerase II p300/CBP associated factor (PCAF) associated factor|TAF5 like RNA polymerase II p300/CBP associated factor associated factor 65 kDa subunit 5L|TAF5 like RNA polymerase II p300/CBP associated factor associated factor 65kDa|TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L|Taf5l|Taf5l protein|TAF5L_HUMAN
- Storage Buffer:PBS, 150 mM NaCl, 0.02% Sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: TAF5L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human