To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Metalloendopeptidase OMA1, mitochondrial protein (22-194AA)
- Antigen Symbol:OMA1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q96E52
- Antigen Synonyms:2010001O09Rik|DAB1|FLJ33782|Metalloendopeptidase Metalloendopeptidase mitochondrial|Metalloprotease-related protein 1|mitochondrial|MPRP 1|MPRP-1|MPRP1|OMA1 homolog zinc metallopeptidase|OMA1 homolog|zinc metallopeptidase (S. cerevisiae)|OMA1 zinc metallopeptidase|OMA1 zinc metallopeptidase homolog|OMA1_HUMAN|Overlapping activity with M-AAA protease|Overlapping with the m-AAA protease 1 homolog|peptidase|YKR087C|Zinc metallopeptidase ZMPOMA1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: OMA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human