To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Troponin I, cardiac muscle protein (2-210AA)
- Antigen Symbol:TNNI3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Size:50 μg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P19429
- Antigen Synonyms:cardiac muscle|Cardiac troponin I|cardiomyopathy|dilated 2A (autosomal recessive)|Cardiomyopathy|familial hypertrophic|7|included|CMD1FF|CMD2A|CMH7|cTnI|Familial hypertrophic cardiomyopathy 7|MGC116817|RCM1|Tn1|Tni|TNN I3|TNNC 1|TNNC1|TNNI3_HUMAN|Troponin I|Troponin I cardiac|Troponin I cardiac muscle|Troponin I cardiac muscle isoform|Troponin I type 3 cardiac|troponin I|cardiac 3|TroponinI|Ttroponin I type 3 (cardiac)
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:200 to 1:500, IP:1:200 to 1:2000
Type:
Antigen: TNNI3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat