Order Entry
United States
ContactUsLinkComponent
Anti-RAF1 Rabbit Polyclonal Antibody
Anti-RAF1 Rabbit Polyclonal Antibody
Catalog # RL612-401-E24
undefined
Anti-RAF1 Rabbit Polyclonal Antibody
Catalog # RL612-401-E24
Supplier Number:  612-401-E24
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Raf-1 Proto-Oncogene, Serine/Threonine Kinase
  • Antigen Symbol:
    RAF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid
  • Gene ID:
    24703
  • Antigen Synonyms:
    Raf-1|CRAF|RAF|NS5|cRaf|C-raf|c-Raf|Proto-oncogene c-RAF
  • Modification:
    phospho-Ser642
  • Storage Buffer:
    0.01 M HEPES, 0.15 M Sodium Chloride, pH 7.50.1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
  • Storage Temperature:
    –20 °C
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-Raf-1 pS642 Antibody was produced in rabbit by repeated immunizations with synthetic phospho-peptide corresponding to amino acid residues surrounding Ser642 conjugated to KLH.
  • Purification:
    affinity-purified
  • Cat. No.:
    RL612-401-E24
  • Supplier no.:
    612-401-E24

Specifications

About this item

Anti-Raf 1 pS642 antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 74 kDa in size corresponding to Raf-1 Protein phosphorylated at Ser642 in the appropriate cell lysate.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat