To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:23'-cyclic nucleotide 3' phosphodiesterase
- Antigen Symbol:CNP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Rabbit,Sheep,Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Antiserum
- Form:Liquid
- Gene ID:25275
- Antigen Synonyms:CNPI|CNPII|Cnp1|CNPF
- Storage Temperature:–20 °C
- Shipping Temperature:Dry Ice
- Immunogen:Anti-CNP Antibody was produced in rabbit by repeated immunizations with endogenous rabbit 2,3 cyclic nucleotide-3-phospho-diesterase
- Cat. No.:RL100-401-D21
- Supplier no.:100-401-D21
Specifications
About this item
Anti-CNP is suitable for usage in ELISA, Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a 46kDa band by western blot corresponding to CNP Protein in the appropriate cell lysate of tissue.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000; Immunochemistry: 1:1000
Type: Primary
Antigen: CNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: