To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Replication factor C subunit 4
- Antigen Symbol:RFC4
- Clonality:Monoclonal
- Clone:1320
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:5984
- Antigen Synonyms:A1|MGC27291|P37|RFC37
- Storage Buffer:Phosphate-buffered saline, pH 7.4, containing no preservatives
- Molecular Weight:37 kDa
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encoding the full-length p37 subunit ofhuman RFC purified from E. coli.
- Tested Applications:WB
- Purification:Protein G Affinity Purified
- Cat. No.:89364-854
- Supplier no.:GTX70285
Specifications
About this item
Mouse monoclonal antibody to human replication factor C ( RFC ) p37
For WB: Use at a dilution of 1:1,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researhcer.
Type: Primary
Antigen: RFC4
Clonality: Monoclonal
Clone: 1320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse