To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:5'-deoxyribose-5-phosphate lyase Ku70
- Antigen Symbol:Ku70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:2547
- Antigen Synonyms:CTC box binding factor 75 kDa subunit|DNA repair protein XRCC6|lupus Ku autoantigen protein p70|thyroid|Ku autoantigen|70 kDa subunit of Ku antigen|Ku autoantigen p70 subunit|70 kDa subunit|ATP-dependent DNA helicase II|5'-dRP lyase Ku70|5'-deoxyribose-5-phosphate lyase Ku70|CTC box-binding factor 75 kDa subunit|70kDa,X-ray repair cross-complementing protein 6|ATP-dependent DNA helicase 2 subunit 1|thyroid autoantigen 70kD (Ku antigen)|ATP-dependent DNA helicase II 70 kDa subunit
- Storage Buffer:Phosphate-buffered saline, pH 7.4, containing no preservatives
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Recombinant antigen containing the amino terminal portion of human Ku70 protein
- Tested Applications:WB
- Purification:Rabbit Polyclonal Antiserum
- Cat. No.:89364-826
- Supplier no.:GTX70259
Rabbit polyclonal antiserum to human Ku70
For WB: Use at a dilution of 1:2,500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: Ku70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human