To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Hemoglobin
- Antigen Symbol:HBA1
- Clonality:Monoclonal
- Clone:901
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3039
- Antigen Synonyms:HBA-T3|HBH
- Storage Buffer:Phosphate-buffered saline, pH 7.4, containing no preservatives
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1.1 mg/ml
- Immunogen:Full length native protein (purified) (Human)
- Tested Applications:ELISA
- Purification:Protein G purified
- Cat. No.:89363-088
- Supplier no.:GTX40079
Mouse monoclonal [901] to Hemoglobin
For ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: HBA1
Clonality: Monoclonal
Clone: 901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human