To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:NAK ATPASE
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Environmentally Preferable:
- Storage Buffer:N/A
- Storage Temperature:Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Purified Na,K-ATPase from rabbit renal outer medulla.
- Tested Applications:WB
- Purification:Ascitic fluid
- Cat. No.:89362-890
- Supplier no.:GTX30203
Specifications
About this item
Na,K-ATPase is an integral membrane protein complex that hydrolyzes ATP to maintain the transmembrane gradients of Na+ and K+ found in most mammalian cells. The enzyme is comprised of an alpha and Beta subunit. The a-polypeptide has been shown to be the catalytically active subunit, whereas the Beta-polypeptide is glycosylated and appears to be necessary for the assembly and transport of the sodium pump to the plasma membrane.
Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human