Order Entry
United States
ContactUsLinkComponent
Anti-CALCA Mouse Monoclonal Antibody [clone: 13f2]
Anti-CALCA Mouse Monoclonal Antibody [clone: 13f2]
Catalog # 89350-918
Supplier:  Genetex
undefined
Anti-CALCA Mouse Monoclonal Antibody [clone: 13f2]
Catalog # 89350-918
Supplier:  Genetex
Supplier Number:  GTX14818
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Calcitonin-related Polypeptide Alpha
  • Antigen Symbol:
    CALCA
  • Clonality:
    Monoclonal
  • Clone:
    13f2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    796
  • Antigen Synonyms:
    CT|Calcitonin-related Polypeptide alpha|CGRP|CALC1|KC|CGRP1|CGRP-I|Calcitonin
  • Storage Buffer:
    Preservative: 0.1% Sodium Azide; Constituents: PBS
  • Storage Temperature:
    Keep as concentrated solution, aliquot and store at 4°C.
  • Concentration:
    2.5 mg/ml
  • Immunogen:
    Calcitonin protein fragment (purified) (Human) (see figure).
  • Tested Applications:
    Enzyme Immunoassay
  • Purification:
    Protein A purified
  • Cat. No.:
    89350-918
  • Supplier no.:
    GTX14818

Specifications

About this item

Calcitonin is a peptide hormone synthesized by the parafollicular cells of the thyroid. It causes a rapid, but short lived, reduction in serum calcium and phosphate by promoting the incorporation of those ions in the bones. This effect is opposite to that of parathyroid hormone.

ELISA: Use at an assay dependent dilution. EIA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 16 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 13f2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human