Order Entry
United States
ContactUsLinkComponent
Anti-MYO6 Mouse Monoclonal Antibody [clone: MUD-19]
Anti-MYO6 Mouse Monoclonal Antibody [clone: MUD-19]
Catalog # 89356-500
Supplier:  Genetex
Anti-MYO6 Mouse Monoclonal Antibody [clone: MUD-19]
Catalog # 89356-500
Supplier:  Genetex
Supplier Number:  GTX11095
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Myosin 6
  • Antigen Symbol:
    MYO6
  • Clonality:
    Monoclonal
  • Clone:
    MUD-19
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 μl
  • Environmentally Preferable:
  • Gene ID:
    4646
  • Antigen Synonyms:
    DFNA 22|MyosinVI|Myo6|DFNB37|DFNB 37|Myosin VI|MYO 6|KIAA0389|Deafness autosomal recessive 37|MYO6_HUMAN|Myosin 6|Unconventional myosin-6.|Myosin-VI|Myosin6|DFNA22
  • Storage Buffer:
    Preservative: 15% Sodium Azide; Constituents: 1% BSA, 0.01M PBS. pH 7.4
  • Storage Temperature:
    Upon delivery aliquot and store at –20 °C. Avoid repeated freeze / thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Synthetic peptide, corresponding to amino acids 291-302 of Human MYO6.
  • Tested Applications:
    ELISA, WB
  • Purification:
    Protein A purified
  • Cat. No.:
    89356-500
  • Supplier no.:
    GTX11095

Specifications

About this item

Mouse monoclonal [MUD - 19] to MYO6

ELISA: Use at an assay dependent concentration. WB: Use at a concentration of 2 ?g/ml. Detects a band of approximately 150 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MYO6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human