Order Entry
United States
ContactUsLinkComponent
Anti-MMP7 Rabbit Polyclonal Antibody
Anti-MMP7 Rabbit Polyclonal Antibody
Catalog # 89350-450
Supplier:  Genetex
undefined
Anti-MMP7 Rabbit Polyclonal Antibody
Catalog # 89350-450
Supplier:  Genetex
Supplier Number:  GTX104658
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Matrix Metalloproteinase 7
  • Antigen Symbol:
    MMP7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    4316
  • Antigen Synonyms:
    PUMP-1|MPSL1|MMP-7
  • Storage Buffer:
    1XPBS, 40% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular Weight:
    30 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MMP7. The exact sequence is proprietary.
  • Tested Applications:
    WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89350-450
  • Supplier no.:
    GTX104658

Specifications

About this item

Rabbit polyclonal antibody to MMP7 (matrix metallopeptidase 7 (matrilysin, uterine))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human