To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Chromogranin A
- Antigen Symbol:CgA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:0.5 ml
- Environmentally Preferable:
- Epitope:L-P-V-N-S-P-M-N-K-G-D-T-E-V-M
- Gene ID:1113
- Antigen Synonyms:betagranin (N-terminal fragment of chromogranin A)|SP-I|chromogranin A (parathyroid secretory protein 1)|chromogranin-A|CGA|Pituitary secretory protein I|parathyroid secretory protein 1
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:5 mg/ml
- Immunogen:Synthetic peptide corresponding to the amino acids 1 - 15 of the native molecule conjugated to Keyhole limpet Haemocyanin.
- Purification:IgG purified
- Cat. No.:102111-716
- Supplier no.:NB100-62464
Specifications
About this item
The Chromogranin A Antibody from Novus Biologicals is a sheep polyclonal antibody to Chromogranin A. This antibody reacts with human. The Chromogranin A Antibody has been validated for the following applications: ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:25 - 1:100
Type: Primary
Antigen: Chromogranin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: L-P-V-N-S-P-M-N-K-G-D-T-E-V-M
Host: Sheep
Isotype: IgG
Reactivity: Human