To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Extracellular link Domain Containing 1
- Antigen Symbol:LYVE-1
- Clonality:Monoclonal
- Clone:RM0033-4D17
- Conjugation:Unconjugated
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:10894
- Antigen Synonyms:lymphatic vessel endothelial hyaluronic acid receptor 1|CRSBP-1|extracellular link domain-containing 1|extracellular link domain containing 1|CRSBP1|HAR|Hyaluronic acid receptor|Extracellular link domain-containing protein 1|LYVE-1XLKD1|cell surface retention sequence binding protein-1|Cell surface retention sequence-binding protein 1|lymphatic vessel endothelial hyaluronan receptor 1
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:Produced from a hybridoma fused by a mouse myeloma with B cells obtained from a rat immunized with mouse LYVE-1 recombinant protein.
- Purification:Protein G purified
- Cat. No.:102124-430
- Supplier no.:NB110-61026
Specifications
About this item
Rat Monoclonal LYVE1 Antibody (RM0033-4D17). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:200
Type: Primary
Antigen: LYVE1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse