To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Pan Macrophage Marker, BM8
- Antigen Symbol:F4/80
- Clonality:Monoclonal
- Clone:CI-A3-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.125 mg
- Environmentally Preferable:
- Gene ID:13733
- Antigen Synonyms:cell surface glycoprotein F4/80|hormone receptor-like sequence 1|EGF-like module containing|Ly71|Gpf480|EGF-like module receptor 1|Rat EMR1|F4/80|EMR1 hormone receptor|TM7LN3|lymphocyte antigen 71|DD7A5-7|mucin-like|Mouse EMR1|EGF-TM7|EGF-like module-containing mucin-like hormone receptor-like 1
- Storage Buffer:Phosphate buffered saline
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Thioglycollate stimulated peritoneal macrophages from C57/BL mice.
- Purification:Protein G purified
- Cat. No.:102133-916
- Supplier no.:NB600-404
The F4 / 80 Antibody (CI-A3-1) from Novus Biologicals is a rat monoclonal antibody to F4 / 80. This antibody reacts with human, mouse. The F4 / 80 Antibody (CI-A3-1) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay, Electron Microscopy.
Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: CI-A3-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse