To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Zyxin
- Antigen Symbol:Zyxin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:7791
- Antigen Synonyms:MYND-type containing 8|PRO2893|predicted protein of HQ2893|Zinc finger MYND domain-containing protein 8|cutaneous T-cell lymphoma associated antigen se14-3|CTCL-associated antigen se14-3|zinc finger|Rack7|MGC31836|KIAA1125|PRKCBP1|protein kinase C-binding protein 1|Cutaneous T-cell lymphoma-associated antigen se14-3|RACK7protein kinase C binding protein 1|zinc finger MYND domain containing protein 8|CTCL tumor antigen se14-3
- Storage Buffer:10mM PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide conjugated by N-terminal added cysteine to KLH, corresponding to amino acids 134-150 of Human Zyxin.
- Purification:IgG purified
- Cat. No.:102133-460
- Supplier no.:NB600-1533
Specifications
About this item
The Zyxin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Zyxin. This antibody reacts with human, mouse, rat. The Zyxin Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Zyxin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat