To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin specific peptidase 15
- Antigen Symbol:USP15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:9958
- Antigen Synonyms:EC 3.1.2.15|ubiquitin thioesterase 15|EC 3.4.19.12|Ubiquitin thiolesterase 15|MGC131982|KIAA0529ubiquitin carboxyl-terminal hydrolase 15|unph-2|MGC149838|Ubiquitin-specific-processing protease 15|ubiquitin specific peptidase 15|UNPH4|ubiquitin specific protease 15|Deubiquitinating enzyme 15|MGC74854
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 960 and the C-terminus of human Ubiquitin Specific Peptidase 15 (AAD41086.1).
- Purification:Immunogen affinity purified
- Cat. No.:102122-012
- Supplier no.:NB110-40690
Specifications
About this item
The USP15 Antibody from Novus Biologicals is a rabbit polyclonal antibody to USP15. This antibody reacts with human, mouse, rat. The USP15 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: USP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat