To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ulex Europaeus Lectin 1
- Antigen Symbol:UEA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Plant,Human
- Size:0.25 ml
- Environmentally Preferable:
- Antigen Synonyms:Gorse or Furze Lectin|Anti H(O) lectin I|UEA-1|H(O) lectin 1
- Storage Buffer:Whole antisera
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Ulex europaeus agglutinin isolated by affinity chromatography from seeds of Ulex europaeus (gorse seed).
- Purification:Unpurified
- Cat. No.:102121-516
- Supplier no.:NB110-13922
Specifications
About this item
The Ulex Europaeus Lectin 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Ulex Europaeus Lectin 1. This antibody reacts with human, plant. The Ulex Europaeus Lectin 1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:500
Type: Primary
Antigen: Ulex Europaeus Lectin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Plant