To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin-conjugating enzyme E2B
- Antigen Symbol:HR6B/UBE2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:7320
- Antigen Synonyms:HR6B|Ubiquitin-conjugating enzyme E2-17 kDa|UBC2|Ubiquitin-protein ligase B|E2 protein|HHR6B|ubiquitin-conjugating enzyme E2B (RAD6 homolog)|E2-17kDa|Ubiquitin carrier protein B|RAD6 homolog B|RAD6BEC 6.3.2.19|ubiquitin-conjugating enzyme E2 B
- Storage Buffer:PBS (pH 7.0), 1.0% BSA and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human UBE2B. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102164-428
- Supplier no.:NBP1-31293
Specifications
About this item
Rabbit Polyclonal Ube2B Antibody. Tested Applications: Western Blot, ELISA. Tested Reactivity: Human.
Type: Primary
Antigen: UBE2B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human