About this item
The TREK 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TREK 1. This antibody reacts with human, mouse, bovine. The TREK 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry (Negative), ICC / IF (Negative).
Type: Primary
Antigen: TREK 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Bovine
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102122-298
- 102122-286
- Antigen Symbol:
- TREK 1
- TREK 1
- Antigen Name:
- Twik-Related K+ Channel 1
- Twik-Related K+ Channel 1
- Antigen Synonyms:
- tandem-pore-domain potassium channel TREK-1|subfamily K|TWIK-related potassium channel 1|MGC126744|two-pore potassium channel 1|TREK-1 K(+) channel subunit|member 2|potassium channel|MGC126742
- tandem-pore-domain potassium channel TREK-1|subfamily K|TWIK-related potassium channel 1|MGC126744|two-pore potassium channel 1|TREK-1 K(+) channel subunit|member 2|potassium channel|MGC126742
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Bovine, Mouse
- Human, Bovine, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 3776
- 3776
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A synthetic peptide made to an N-terminal region of human TREK 1 (within residues 1-100). [Swiss-Prot# Q9NRT2]
- A synthetic peptide made to an N-terminal region of human TREK 1 (within residues 1-100). [Swiss-Prot# Q9NRT2]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Tris-Glycine and 0.15M NaCl
- Tris-Glycine and 0.15M NaCl
- Storage Temperature:
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.