To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Toll-like Receptor 4
- Antigen Symbol:TLR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:7099
- Antigen Synonyms:toll-like receptor 4|CD284 antigen|CD284|ARMD10|TOLL|hTollhomolog of Drosophila toll
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:A portion of amino acids 100-150 of human TLR4 was used as the immunogen.
- Purification:Protein G purified
- Cat. No.:102110-136
- Supplier no.:NB100-56579
Specifications
About this item
The TLR4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR4. This antibody reacts with human, mouse. The TLR4 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: TLR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse