To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Tetraspanin-4
- Antigen Symbol:TSPAN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:7106
- Antigen Synonyms:TSPAN-4|TETRASPAN|tetraspanin 4|tetraspanin-4|Novel antigen 2|Transmembrane 4 superfamily member 7NAG-2NAG2TM4SF7tetraspan TM4SF
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to TSPAN4 (tetraspanin 4) The peptide sequence was selected from the middle region of TSPAN4) Peptide sequence YTDKIDRYAQQDLKKGLHLYGTQGNVGLTNAWSIIQTDFRCCGVSNYTDW (NP_001020405).
- Purification:Immunogen affinity purified
- Cat. No.:102192-568
- Supplier no.:NBP1-59438
Specifications
About this item
The Tetraspanin-4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Tetraspanin-4. This antibody reacts with human. The Tetraspanin-4 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.
Type: Primary
Antigen: Tetraspanin-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human