To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Transcription Factor 12
- Antigen Symbol:TCF-12/HTF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:6938
- Antigen Synonyms:transcription factor 12|Transcription factor HTF-4|bHLHb20E-box-binding protein|HEBDNA-binding protein HTF4|BHLHB20|Class B basic helix-loop-helix protein 20|helix-loop-helix transcription factor 4|HsT17266|HTF4TCF-12
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TCF12. The immunogen is located within amino acids 50 - 100 of TCF12.
- Purification:Immunogen affinity purified
- Cat. No.:102207-502
- Supplier no.:NBP1-76304
Specifications
About this item
The TCF-12 / HTF4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TCF-12 / HTF4. This antibody reacts with human, mouse, rat. The TCF-12 / HTF4 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: TCF-12/HTF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat