To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Epimorphin/Syntaxin 2
- Antigen Symbol:STX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse,Hamster,Drosophila,Pig,Rabbit,Monkey,Xenopus,Sheep,Chicken,Human,Bovine,Dog
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2054
- Antigen Synonyms:STX2Bsyntaxin-2|syntaxin 2|STX2CEpimorphin|STX2AMGC51014|EPIMepimorphin|EPM
- Storage Buffer:PBS (pH 7.2) and 50% Glycerol
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic peptide derived from sequence near the amino-terminus of rat Syntaxin 2, conjugated to KLH
- Purification:Immunogen affinity purified
- Cat. No.:102158-688
- Supplier no.:NBP1-19338
Specifications
About this item
The Epimorphin / Syntaxin 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Epimorphin / Syntaxin 2. This antibody reacts with human, mouse, rat, porcine, bovine, canine, chicken, drosophila, hamster, monkey, rabbit, sheep, xenopus. The Epimorphin / Syntaxin 2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Epimorphin/Syntaxin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Hamster, Monkey, Rabbit, Sheep, Xenopus