Order Entry
United States
ContactUsLinkComponent
 
Anti-Synaptogyrin 3 Rabbit Polyclonal Antibody
  102210-598
Anti-Synaptogyrin 3 Rabbit Polyclonal Antibody
  102210-598
 :  NBP1-77102
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Synaptogyrin 3
  • Antigen Symbol:
    SYNGR3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    9143
  • Antigen Synonyms:
    synaptogyrin-3|synaptogyrin 3|MGC20003
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human SYNGR3. The immunogen is located within the last 50 amino acids of SYNGR3.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102210-598
  • Supplier no.:
    NBP1-77102

 

 

The Synaptogyrin 3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Synaptogyrin 3. This antibody reacts with human, mouse, rat. The Synaptogyrin 3 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: Synaptogyrin 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat