Order Entry
United States
ContactUsLinkComponent
Anti-SSRP1 Rabbit Polyclonal Antibody
Anti-SSRP1 Rabbit Polyclonal Antibody
  102166-874
Anti-SSRP1 Rabbit Polyclonal Antibody
  102166-874
 :  NBP1-33235
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Structure Specific Recognition Protein 1
  • Antigen Symbol:
    SSRP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    6749
  • Antigen Synonyms:
    FACT complex subunit SSRP1|Facilitates chromatin transcription complex subunit SSRP1|Chromatin-specific transcription elongation factor 80 kDa subunit|Structure-specific recognition protein 1|FACT|FACTp80|Facilitates chromatin transcription complex 80 kDa subunit|high mobility group box|facilitates chromatin remodeling 80 kDa subunit|structure specific recognition protein 1|cisplatin-DNA SSRP|Recombination signal sequence recognition protein 1|T160|FACT80FACT 80 kDa subunit|hSSRP1
  • Storage Buffer:
    0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human SSRP1. The exact sequence is proprietary.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102166-874
  • Supplier no.:
    NBP1-33235

 

 

The SSRP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SSRP1. This antibody reacts with human. The SSRP1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SSRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human