Order Entry
United States
ContactUsLinkComponent
Anti-SRPX2 Rabbit Polyclonal Antibody
Anti-SRPX2 Rabbit Polyclonal Antibody
  102211-652
Anti-SRPX2 Rabbit Polyclonal Antibody
  102211-652
 :  NBP1-77370
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Sushi-Repeat Containing Protein, X-Linked 2
  • Antigen Symbol:
    SRPX2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    27286
  • Antigen Synonyms:
    RESDX|sushi-repeat-containing protein|CBPS|X-linked 2|sushi-repeat containing protein|sushi-repeat protein up-regulated in leukemia|sushi repeat-containing protein SRPX2|SRPULPMGX|BPP
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human SRPX2. The immunogen is located within the first 50 amino acids of SRPX2.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102211-652
  • Supplier no.:
    NBP1-77370

 

 

The SRPX2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRPX2. This antibody reacts with human, mouse, rat. The SRPX2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SRPX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat