About this item
The SLC31A1 / CTR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC31A1 / CTR1. This antibody reacts with human, mouse, rat, porcine, xenopus, zebrafish. The SLC31A1 / CTR1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:250
Type: Primary
Antigen: SLC31A1/CTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Xenopus, Zebrafish
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102108-830
- 102108-818
- Antigen Symbol:
- SLC31A1/CTR1
- SLC31A1/CTR1
- Antigen Name:
- Solute Carrier Family 31 Member A1
- Solute Carrier Family 31 Member A1
- Antigen Synonyms:
- solute carrier family 31 (copper transporters)|Solute carrier family 31 member 1|CTR1MGC75487|Copper transporter 1|hCTR1copper transport 1 homolog|COPT1high affinity copper uptake protein 1|member 1
- solute carrier family 31 (copper transporters)|Solute carrier family 31 member 1|CTR1MGC75487|Copper transporter 1|hCTR1copper transport 1 homolog|COPT1high affinity copper uptake protein 1|member 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Zebrafish, Pig, Xenopus, Human, Rat, Mouse
- Zebrafish, Pig, Xenopus, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1317
- 1317
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A synthetic peptide derived from a C-terminal sequence of human SLC31A1/CTR1 [UniProt# O15431]
- A synthetic peptide derived from a C-terminal sequence of human SLC31A1/CTR1 [UniProt# O15431]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:
- Store at 4C. Do not freeze.
- Store at 4C. Do not freeze.