To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Ribosomal Protein L22
- Antigen Symbol:RPL22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:6146
- Antigen Synonyms:HBP15/L22|heparin-binding protein 15|Epstein-Barr-encoded RNA-associated protein|Heparin-binding protein HBp15|EAPHBP15|ribosomal protein L22,60S ribosomal protein L22|EBER-associated protein|Epstein-Barr virus small RNA-associated protein
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide corresponding to aa 79-128, from the C-terminal region of mouse RPL22 (NP_033105). The peptide is homologous in human. Peptide sequence SKRYLKYLTKKYLKKNNLRDWLRVVANSKESYELRYFQINQDEEEEEDED.
- Purification:Immunogen affinity purified
- Cat. No.:102223-208
- Supplier no.:NBP1-98446
The RPL22 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RPL22. This antibody reacts with mouse. The RPL22 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: RPL22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse