To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Ran-binding Protein 2
- Antigen Symbol:RanBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:5903
- Antigen Synonyms:RanBP2|P270|RAN binding protein 2|EC 5.2.1.8|ANE1|nucleoporin 358|Nuclear pore complex protein Nup358|E3 SUMO-protein ligase RanBP2|NUP358TRP2|TRP1|Ran-binding protein 2|transformation-related protein 2,358 kDa nucleoporin|Nucleoporin Nup358
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1050 and 1100 of human RAN binding protein 2 using the numbering given in entry NP_006258.3 (GeneID 5903).
- Purification:Immunogen affinity purified
- Cat. No.:102120-794
- Supplier no.:NB100-93337
The RanBP2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RanBP2. This antibody reacts with human, mouse. The RanBP2 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: RanBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse