To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:RAs-related Nuclear protein
- Antigen Symbol:Ran
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:5901
- Antigen Synonyms:RanGTPase|ARA24Gsp1|TC4|OK/SW-cl.81|Ras-related nuclear protein|GTP-binding nuclear protein Ran|RAN|guanosine triphosphatase Ran|GTPase Ran|Androgen receptor-associated protein 24|member RAS oncogene family|Ras-like protein TC4
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:0.6 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human RAN. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102165-092
- Supplier no.:NBP1-31776
Specifications
About this item
The Ran Antibody from Novus Biologicals is a rabbit polyclonal antibody to Ran. This antibody reacts with human, mouse. The Ran Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Ran
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse