To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Phosphoribosyl Pyrophosphate Synthetase 2
- Antigen Symbol:PRPS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100μl
- Environmentally Preferable:
- Gene ID:5634
- Antigen Synonyms:PRSII|phosphoribosyl pyrophosphate synthetase 2|Phosphoribosyl pyrophosphate synthase II|EC 2.7.6.1|ribose-phosphate diphosphokinase 2|PPRibP synthetase|PPRibP|PRS-II|ribose-phosphate pyrophosphokinase 2
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to PRPS2(phosphoribosyl pyrophosphate synthetase 2) The peptide sequence was selected from the N terminal of PRPS2. Peptide sequence MPNIVLFSGSSHQDLSQRVADRLGLELGKVVTKKFSNQETSVEIGESVRG.
- Purification:Protein A purified
- Cat. No.:102188-254
- Supplier no.:NBP1-56666
Specifications
About this item
The PRPS2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PRPS2. This antibody reacts with human. The PRPS2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: PRPS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human