To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Protein L-isoaspartyl methyltransferase
- Antigen Symbol:PIMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:96764
- Antigen Synonyms:PRIP-interacting protein PIPMT|Hepatocellular carcinoma-associated antigen 137|EC 2.1.1.-|HCA137|PRIP-interacting protein with methyltransferase motif|nuclear receptor coactivator 6 interacting protein|trimethylguanosine synthase homolog|EC 2.1.1|PIPMT|trimethylguanosine synthase 1|PIMTFLJ22995|trimethylguanosine synthase|CLL-associated antigen KW-2|DKFZp762A163|PRIP-interacting protein with methyltransferase domain|Nuclear receptor coactivator 6-interacting protein|trimethylguanosine synthase homolog (S. cerevisiae)|NCOA6IPSEREX-defined|Cap-specific guanine-N2 methyltransferase
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to TGS1(trimethylguanosine synthase homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of TGS1. Peptide sequence IDENPASDFDDSGSLLGFKYGSGQKYGGIPNFSHRQVRYLEKNVKLKSKY.
- Purification:Immunogen affinity purified
- Cat. No.:102184-840
- Supplier no.:NBP1-54942
Specifications
About this item
The PIMT Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIMT. This antibody reacts with human. The PIMT Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: PIMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human