To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Protein inhibitor of activated STAT protein 4
- Antigen Symbol:PIASy
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:51588
- Antigen Synonyms:E3 SUMO-protein ligase PIAS4|ZMIZ6|PIAS-gamma|zinc finger|protein inhibitor of activated STAT protein PIASy|MGC35296|4|Piasg|MIZ-type containing 6|Protein inhibitor of activated STAT protein 4|protein inhibitor of activated STAT|FLJ12419|PIASy|Protein inhibitor of activated STAT protein gamma
- Storage Buffer:PBS containing 0.05% BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:This polyclonal antibody was developed against a mixture of synthetic peptides corresponding to amino acids 46-60 (KKIKELYETRYAKKNC) and 187-198 (YSDTSCPQEDQY) of human PIASg; GenBank no. AAD45155.1 AF164437_1. These sequences are 95% and 100% homologous in mouse and rat.
- Purification:Protein G purified
- Cat. No.:102133-192
- Supplier no.:NB600-1318
Specifications
About this item
The PIASy Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIASy. This antibody reacts with human, mouse, rat. The PIASy Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: PIASy
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat