To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Homeobox protein NANOG
- Antigen Symbol:Nanog
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:79923
- Antigen Synonyms:homeobox protein NANOG|hNanog|Nanog homeobox|FLJ40451|homeobox transcription factor Nanog-delta 48|Homeobox transcription factor Nanog|FLJ12581
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:A synthetic peptide made to the mouse Nanog protein (within residues 1-50). [Swiss-Prot Q80Z64]
- Purification:Immunogen affinity purified
- Cat. No.:102110-672
- Supplier no.:NB100-58842
Rabbit Polyclonal Nanog Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500
Type: Primary
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse