Order Entry
United States
ContactUsLinkComponent
Anti-MAP8 Rabbit Polyclonal Antibody
Anti-MAP8 Rabbit Polyclonal Antibody
Catalog # 102176-976
Supplier:  Novus Biologicals
Anti-MAP8 Rabbit Polyclonal Antibody
Catalog # 102176-976
Supplier:  Novus Biologicals
Supplier Number:  NBP1-47281
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    MAP1S
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    55201
  • Antigen Synonyms:
    C19orf5|VCY2 interacting protein 1|MAP8MAP-1S|Microtubule-associated protein 8|FLJ10669|Variable charge Y chromosome 2-interacting protein 1|microtubule-associated protein 1S|VCY2IP1BPY2-interacting protein 1|chromosome 19 open reading frame 5|BPY2 interacting protein 1|VCY2-interacting protein 1|BPY2IP1VCY2IP-1|MGC133087
  • Storage Buffer:
    TBS and 0.1% BSA
  • Storage Temperature:
    Store at 4C. Do not freeze.
  • Concentration:
    0.2 mg/ml
  • Immunogen:
    The immunogen for this product maps to a region between residue 475 and 525 of human MAP1S using the numbering given in entry NP_060644.4 (GeneID 55201).
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102176-976
  • Supplier no.:
    NBP1-47281

Specifications

About this item

Rabbit Polyclonal MAP8 Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: MAP8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human