To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:KMT3B/NSD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:64324
- Antigen Synonyms:Lysine N-methyltransferase 3B|FLJ22263|ARA267STO|FLJ10684|DKFZp666C163|KMT3BSotos syndrome|H3 lysine-36 and H4 lysine-20 specific|FLJ44628|EC 2.1.1.43|NR-binding SET domain-containing protein|nuclear receptor binding SET domain protein 1|Androgen receptor-associated protein of 267 kDa|Androgen receptor coactivator 267 kDa protein
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 650 and 700 of human NSD1 using the numbering given in SwissProt entry Q96L73 (GeneID 64324).
- Purification:Immunogen affinity purified
- Cat. No.:102105-866
- Supplier no.:NB100-1744
Specifications
About this item
Rabbit Polyclonal KMT3B/NSD1 Antibody. Tested Applications: Immunohistochemistry. Tested Reactivity: Human.
Type: Primary
Antigen: KMT3B/NSD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human