To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Kelch-like 8
- Antigen Symbol:KLHL8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:57563
- Antigen Synonyms:kelch-like 8 (Drosophila)|KIAA1378FLJ46304|kelch-like protein 8
- Storage Buffer:PBS & 2% Sucrose.
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide directed towards the N terminal of human KLHL8. Peptide sequence MASDSMSSKQARNHITKGKRQQQHQQIKNRSSISDGDGEDSFIFEANEAW.
- Purification:Immunogen affinity purified
- Cat. No.:102216-340
- Supplier no.:NBP1-80346
Specifications
About this item
Rabbit Polyclonal KLHL8 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Rat, Bovine, Canine, Drosophila.
Type: Primary
Antigen: KLHL8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human