To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Kinesin Family member 1B
- Antigen Symbol:KIF1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:23095
- Antigen Synonyms:KIAA0591FLJ23699|CMT2A1|Klp|CMT2A|kinesin superfamily protein KIF1B|kinesin-like protein KIF1B|CMT2|NBLST1|kinesin family member 1B|HMSNII|MGC134844|KIAA1448
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1766 and the C-terminus (residue 1816) of human kinesin family member 1B using the numbering given in entry CAC16629.1 (GeneID 23095)
- Purification:Immunogen affinity purified
- Cat. No.:102110-434
- Supplier no.:NB100-57494
Specifications
About this item
Rabbit Polyclonal KIF1B Antibody. Tested Applications: Immunoprecipitation. Tested Reactivity: Human.
Type: Primary
Antigen: KIF1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human