To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:KH Domain Containing RNA binding signal transduction associated 2
- Antigen Symbol:KHDRBS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:202559
- Antigen Synonyms:RNA-binding|KH domain containing|signal transduction associated 2|SLM-1FLJ38664|signal transduction-associated protein 2|hSLM-1|SLM1bA535F17.1|Sam68-like mammalian protein 1|RNA binding|MGC26664|KH domain-containing
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to KHDRBS2(KH domain containing, RNA binding, signal transduction associated 2) The peptide sequence was selected from the middle region of KHDRBS2. Peptide sequence EAYSRMSHALEEIKKFLVPDYNDEIRQEQLRELSYLNGSE
- Purification:Immunogen affinity purified
- Cat. No.:102187-566
- Supplier no.:NBP1-56317
Specifications
About this item
Rabbit Polyclonal KHDRBS2 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: KHDRBS2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human