About this item
Rabbit Polyclonal HP1 alpha Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.
Type: Primary
Antigen: HP1 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102107-556
- 102107-568
- Antigen Symbol:
- HP1A
- HP1A
- Antigen Name:
- HP1 alpha
- HP1 alpha
- Antigen Synonyms:
- chromobox homolog 5 (Drosophila HP1 alpha)|heterochromatin protein 1-alpha|chromobox homolog 5|HP1A|HP1Hs alpha|HP1 alpha|HP1 alpha homolog|HP1|Antigen p25|Drosophila)|chromobox protein homolog 5|HP1Hs-alpha|Heterochromatin protein 1 homolog alpha|chromobox homolog 5 (HP1 alpha homolog|HP1-ALPHA
- chromobox homolog 5 (Drosophila HP1 alpha)|heterochromatin protein 1-alpha|chromobox homolog 5|HP1A|HP1Hs alpha|HP1 alpha|HP1 alpha homolog|HP1|Antigen p25|Drosophila)|chromobox protein homolog 5|HP1Hs-alpha|Heterochromatin protein 1 homolog alpha|chromobox homolog 5 (HP1 alpha homolog|HP1-ALPHA
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 23468
- 23468
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- A synthetic peptide containing an N-terminal region of human HP1 alpha (within residues 1-100). [Swiss-Prot P45973]
- A synthetic peptide containing an N-terminal region of human HP1 alpha (within residues 1-100). [Swiss-Prot P45973]
- Purification:
- IgG purified
- IgG purified
- Storage Buffer:
- PBS
- PBS
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.