To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:High-mobility Group Nucleosome Binding Domain 1
- Antigen Symbol:HMGN1/HMG14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Sheep,Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3150
- Antigen Synonyms:MGC117425|high-mobility group (nonhistone chromosomal) protein 14|High mobility group nucleosome-binding domain-containing protein 1|FLJ31471|MGC104230|non-histone chromosomal protein HMG-14|high-mobility group nucleosome binding 1|nonhistone chromosomal protein HMG-14|HMG14FLJ27265|high-mobility group nucleosome binding domain 1
- Modification:p Ser20, p Ser24
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1.2 mg/ml
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 19-28 of human HMGN protein.
- Purification:Immunogen affinity purified
- Cat. No.:102212-498
- Supplier no.:NBP1-78102
Specifications
About this item
Rabbit Polyclonal HMG14 [p Ser24] Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry. Tested Reactivity: Human.
Type: Primary
Antigen: HMG14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human