To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GRF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:2889
- Antigen Synonyms:Guanine nucleotide-releasing factor 2|DKFZp781P1719|CRK SH3-binding GNRP|C3GGRF2guanine nucleotide-releasing factor 2 (specific for crk proto-oncogene)|rap guanine nucleotide exchange factor 1|Protein C3G|Rap guanine nucleotide exchange factor (GEF) 1
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human CRK SH3-binding GNRP using the numbering given in entry NP_005303.2 (GeneID 2889).
- Purification:Immunogen affinity purified
- Cat. No.:102157-176
- Supplier no.:NBP1-06500
Specifications
About this item
Rabbit Polyclonal GRF2 Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.
Type: Primary
Antigen: GRF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human