To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Fanconi Anemia Complementation Group D2
- Antigen Symbol:FANCD2
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Primates,Human,Dog,Mouse
- Western Blot:Yes
- Size:0.05 ml
- Environmentally Preferable:
- Gene ID:2177
- Antigen Synonyms:Fanconi anemia group D2 protein|FA4|FACDFLJ23826|FANCD|Protein FACD2|Fanconi anemia|DKFZp762A223|FADFAD2|FA-D2|complementation group D2
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
- Purification:Immunogen affinity purified
- Cat. No.:102106-104
- Supplier no.:NB100-182R
Specifications
About this item
The FANCD2 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to FANCD2. This antibody reacts with human, mouse, canine, primate. The FANCD2 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FANCD2
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Canine, Primate