To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Endothelial Lipase
- Antigen Symbol:Endothelial Lipase
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:9388
- Antigen Synonyms:EC 3.1.1|endothelial|EC 3.1.1.3|Endothelial cell-derived lipase|EDLendothelial lipase|ELlipoprotein lipase H|lipase|PRO719
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A C-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9]
- Purification:Immunogen affinity purified
- Cat. No.:102131-372
- Supplier no.:NB400-111G
Specifications
About this item
The Endothelial Lipase Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to Endothelial Lipase. This antibody reacts with human, mouse (negative). The Endothelial Lipase Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.
Type: Primary
Antigen: Endothelial Lipase
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse (Negative)