About this item
The Derlin 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Derlin 1. This antibody reacts with human, mouse, canine, chinese hamster. The Derlin 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Canine, Chinese Hamster
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102109-210
- 102109-196
- Antigen Symbol:
- DER1
- DER1
- Antigen Name:
- Derlin 1
- Derlin 1
- Antigen Synonyms:
- Der1-like protein 1|DER-1|Der1-like domain family|FLJ42092|MGC3067|member 1|PRO2577|FLJ13784|DERtrin-1|derlin-1|DER1Degradation in endoplasmic reticulum protein 1
- Der1-like protein 1|DER-1|Der1-like domain family|FLJ42092|MGC3067|member 1|PRO2577|FLJ13784|DERtrin-1|derlin-1|DER1Degradation in endoplasmic reticulum protein 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Chinese hamster, Human, Dog, Mouse
- Chinese hamster, Human, Dog, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 79139
- 79139
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A synthetic peptide made to the C-terminal region of human Derlin 1 (between residues 200-251). [UniProt# Q9BUN8]
- A synthetic peptide made to the C-terminal region of human Derlin 1 (between residues 200-251). [UniProt# Q9BUN8]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:
- Store at 4C. Do not freeze.
- Store at 4C. Do not freeze.