To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Dynactin Subunit 1
- Antigen Symbol:DCTN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:1639
- Antigen Synonyms:dynactin subunit 1|Glued (Drosophila) homolog)|p135|dynactin 1 (p150|Drosophila)|p150 Glued (Drosophila) homolog|p150-glued|glued homolog|dynactin 1
- Storage Buffer:PBS (pH 7.0), 1.0% BSA and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human DCTN1. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102163-828
- Supplier no.:NBP1-30880
Specifications
About this item
The Dynactin Subunit 1 / DCTN1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Dynactin Subunit 1 / DCTN1. This antibody reacts with human, rat. The Dynactin Subunit 1 / DCTN1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: DCTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat